Methyl-4-iodobenzoate 98%

Supplier: Thermo Scientific Chemicals

41446-0250 41446-0050 41446-1000
TS41446-0250EA 106 USD
TS41446-0250 TS41446-0050 TS41446-1000
Methyl-4-iodobenzoate 98%
Methyl-4-iodobenzoate
Formula: C₈H₇IO₂
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00016353
CAS Number: 619-44-3
EINECS: 210-597-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to beige to light orange Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 113 to 117 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR