Mercury(II) sulphate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury sulphate, Mercuric sulphate

41365-1000 41365-0025 41365-5000
TS41365-1000EA 150 USD
TS41365-1000 TS41365-0025 TS41365-5000
Mercury(II) sulphate ACS
Mercury(II) sulphate
Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850
Density: 6.47 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1 II
Merck Index: 13,05914

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Titration with NH4SCN ≥98.0 %
Chloride (Cl) ≤30 ppm
Nitrate (NO3) Passes test (limit about 0.003%)
Iron (Fe) ≤50 ppm
Mercurous Mercury (as Hg) ≤0.15 %
Residue after reduction ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR