4-Iodobenzonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41238-0050
41238-0250
TS41238-0050EA
104
USD
TS41238-0050
TS41238-0250
4-Iodobenzonitrile 99%
4-Iodobenzonitrile
Formula:
C₇H₄IN Boiling Pt: 285 °C (1013 hPa) Melting Pt: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051310 CAS Number: 3058-39-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | Light yellow to yellow-brown Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 125 to 128 °C |
GC | ≥98.5 % |
Stabilizer | 50 to 150 ppm Amylene |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...