4-Chlorophenylhydrazinium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chlorophenylhydrazine hydrochloride, 1-(4-Chlorophenyl)hydrazine hydrochloride

40495-0050 40495-0250 40495-1000
TS40495-0050EA 36 USD
TS40495-0050 TS40495-0250 TS40495-1000
4-Chlorophenylhydrazinium chloride 97%
4-Chlorophenylhydrazinium chloride
Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
Melting Pt: 203 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012943
CAS Number: 1073-70-7
EINECS: 214-030-9

Order Now

Specification Test Results

Appearance White to pink to light brown Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 96.0 to 104.0 %
Loss on drying ≤3 % (1 g, 105°C)
HPLC ≥96.0 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR