Butyl-4-hydroxybenzoate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

40357-1000 40357-5000
TS40357-1000EA 46 USD
TS40357-1000 TS40357-5000
Butyl-4-hydroxybenzoate 99+%
Butyl-4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
Boiling Pt: 156 °C (3.5 mmHg)
Melting Pt: 68…71 °C
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
Merck Index: 13,01583

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 68 to 71 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR