Bis(2-ethylhexyl)terephthalate 97%

Supplier: Thermo Scientific Chemicals

40249-2500 40249-0010 40249-0100
TS40249-2500EA 25 USD
TS40249-2500 TS40249-0010 TS40249-0100
Bis(2-ethylhexyl)terephthalate 97%
Bis(2-ethylhexyl)terephthalate
Formula: C₂₄H₃₈O₄
Boiling Pt: 400 °C (760 mmHg)
Melting Pt: –67 °C
Density: 0.984 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00072256
CAS Number: 6422-86-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4920 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR