8-Anilino-1-naphthalenesulphonic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid

40121-0250 40121-0050 40121-1000
TS40121-0250EA 173 USD
TS40121-0250 TS40121-0050 TS40121-1000
8-Anilino-1-naphthalenesulphonic acid 98%
8-Anilino-1-naphthalenesulphonic acid
Formula: C₁₆H₁₃NO₃S
Melting Pt: 183…183.5 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003998
CAS Number: 82-76-8

Order Now

Specification Test Results

Appearance Dark green to brown to black Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR