4-Chloro-2-methoxybenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Chloro-2-methoxyphenyl)methanol

39856-0010 39856-0050
TS39856-0010EA 36 USD
TS39856-0010 TS39856-0050
4-Chloro-2-methoxybenzyl alcohol 97%
4-Chloro-2-methoxybenzyl alcohol
Formula: C₈H₉ClO₂
Melting Pt: 47 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD02683547
CAS Number: 55685-75-1

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 47 to 51 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR