(2-Iodophenyl)acetic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(2-Iodophenyl)acetic acid

39404-0010 39404-0050 39404-0250
TS39404-0010EA 26 USD
TS39404-0010 TS39404-0050 TS39404-0250
(2-Iodophenyl)acetic acid 97+%
(2-Iodophenyl)acetic acid
Formula: (C₆H₄I)CH₂COOH
MW: 262.05 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00046546
CAS Number: 18698-96-9

Order Now

Specification Test Results

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 113 to 118 °C
HPLC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR