2-Bromo-α-methylbenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Bromophenyl)ethanol

39364-0010 39364-0050 39364-0250
TS39364-0010EA 17 USD
TS39364-0010 TS39364-0050 TS39364-0250
2-Bromo-α-methylbenzyl alcohol 99%
2-Bromo-α-methylbenzyl alcohol
Formula: C₈H₉BrO
MW: 201.06 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00065001
CAS Number: 5411-56-3
EINECS: 226-491-3

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5610 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR