2-(4-Bromophenyl)-1,3-dioxolane 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromobenzaldehyde Ethylene Acetal

39322-0010 39322-0050
TS39322-0010EA 69 USD
TS39322-0010 TS39322-0050
2-(4-Bromophenyl)-1,3-dioxolane 98+%
2-(4-Bromophenyl)-1,3-dioxolane
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01318950
CAS Number: 10602-01-4

Order Now

Specification Test Results

Appearance Colorless Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5650 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR