3-Methoxybenzyl bromide 98+%

Supplier: Thermo Scientific Chemicals
Danger

39311-0050 39311-0250
TS39311-0050EA 70 USD
TS39311-0050 TS39311-0250
3-Methoxybenzyl bromide 98+%
3-Methoxybenzyl bromide
Formula: C₈H₉BrO
Boiling Pt: 152 °C (760 mmHg)
Density: 1.436 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00216590
CAS Number: 874-98-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR