α-Bromo-4-chlorotoulene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide

39171-0050 39171-0250 39171-1000
TS39171-0050EA 57 USD
TS39171-0050 TS39171-0250 TS39171-1000
α-Bromo-4-chlorotoulene 98%
α-Bromo-4-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 118…123 °C (19 mmHg)
Melting Pt: 46…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00040714
CAS Number: 622-95-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to almost white Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 46 to 51 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR