4-Aminobenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Aminomethyl)aniline, alpha-Amino-p-toluidine

39145-0050 39145-0250 39145-1000
TS39145-0050EA 55 USD
TS39145-0050 TS39145-0250 TS39145-1000
4-Aminobenzylamine 98%
4-Aminobenzylamine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075513
CAS Number: 4403-71-8
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to light brown Liquid or solid
Infrared spectrum Conforms
Refractive index 1.6080 to 1.6120 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR