(4-Chlorophenyl)acetic acid ethyl ester 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-chlorophenylacetate, Ethyl (4-chlorophenyl)acetate

38884-0250 38884-0050
TS38884-0250EA 61 USD
TS38884-0250 TS38884-0050
(4-Chlorophenyl)acetic acid ethyl ester 98+%
(4-Chlorophenyl)acetic acid ethyl ester
Formula: (C₆H₄Cl)CH₂COOCH₂CH₃
MW: 198.65 g/mol
MDL Number: MFCD00018791
CAS Number: 14062-24-9
EINECS: 237-906-2

Order Now

Specification Test Results

Appearance Colorless Low melting solid
Infrared spectrum Conforms
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR