Benzylideneacetone 98+%

Supplier: Thermo Scientific Chemicals
Warning

38559-2500 38559-0010 38559-0500
TS38559-2500EA 36 USD
TS38559-2500 TS38559-0010 TS38559-0500
Benzylideneacetone 98+%
Benzylideneacetone
Formula: C₁₀H₁₀O
Boiling Pt: 261 °C (760 mmHg)
Melting Pt: 38.5 °C
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00008779
CAS Number: 122-57-6

Order Now

Specification Test Results

Appearance Yellow Low melting solid
Infrared spectrum Conforms
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR