(R)-(+)-3-Bromo-α-methylbenzyl alcohol 95%, 98% ee

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-3-Bromo-α-methylbenzyl alcohol, (R)-3-Bromo-alpha-methylbenzyl Alcohol

37649-0010 37649-0050
TS37649-0010EA 106 USD
TS37649-0010 TS37649-0050
(R)-(+)-3-Bromo-α-methylbenzyl alcohol 95%, 98% ee
(R)-(+)-3-Bromo-α-methylbenzyl alcohol
Formula: C₈H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD06658929
CAS Number: 134615-24-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR