α-Phenylbenzyl bromide 90%

Supplier: Thermo Scientific Chemicals
Danger

37346-1000 37346-5000
TS37346-1000EA 45 USD
TS37346-1000 TS37346-5000
α-Phenylbenzyl bromide 90%
α-Phenylbenzyl bromide
Formula: (C₆H₅)₂CHBr
Boiling Pt: 184 °C (20 mmHg)
Melting Pt: 33…42 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000134
CAS Number: 776-74-9
EINECS: 212-279-8
UN: 1770
ADR: 8,II

Order Now

Specification Test Results

Appearance Orange to brown Low melting crystalline mass or solid
Infrared spectrum Conforms
Melting point 33 to 42 °C
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR