1,2,4,5-Tetrakis(bromomethyl)benzene 95%

Supplier: Thermo Scientific Chemicals
Danger

37092-0050 37092-0250
TS37092-0050EA 178 USD
TS37092-0050 TS37092-0250
1,2,4,5-Tetrakis(bromomethyl)benzene 95%
1,2,4,5-Tetrakis(bromomethyl)benzene
Formula: C₁₀H₁₀Br₄
MW: 449.8 g/mol
Melting Pt: 156…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00009664
CAS Number: 15442-91-8
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 155 to 161 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR