2-Thiopheneethanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(2-Thienyl)ethanol, Thiophen-2-ethanol

36654-0250 36654-1000
TS36654-0250EA 47 USD
TS36654-0250 TS36654-1000
2-Thiopheneethanol 98%
2-Thiopheneethanol
Formula: C₆H₈OS
MW: 128.19 g/mol
Boiling Pt: 108 °C (13 mmHg)
Density: 1.153 g/cm³
Flash Pt: 101 °C
MDL Number: MFCD00005462
CAS Number: 5402-55-1
EINECS: 226-452-0

Order Now

Specification Test Results

Appearance Clear light yellow to grey-green to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5490 to 1.5520 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR