4-Iodophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Iodophenyl)boronic acid, Dihydroxy(4-iodophenyl)borane, 4-Iodobenzeneboronic acid, (4-Iodophenyl)metaboric acid

35948-0010 35948-0050 35948-0250
TS35948-0010EA 38 USD
TS35948-0010 TS35948-0050 TS35948-0250
4-Iodophenylboronic acid 97%
4-Iodophenylboronic acid
Formula: C₆H₆BIO₂
MW: 247.83 g/mol
Melting Pt: 322…326 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01319014
CAS Number: 5122-99-6

Order Now

Specification Test Results

Appearance Light beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR