2,4-Difluorophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

35913-0010 35913-0100 35913-0500
TS35913-0010EA 23 USD
TS35913-0010 TS35913-0100 TS35913-0500
2,4-Difluorophenylboronic acid 98%
2,4-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
Melting Pt: 247…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318998
CAS Number: 144025-03-6

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder or crystals
Infrared spectrum Conforms
Melting point 247 to 251 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR