Dipentyl phthalate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Di-n-pentyl phthalate
35736-0250
35736-1000
35736-5001
TS35736-0250EA
217
USD
TS35736-0250
TS35736-1000
TS35736-5001
Dipentyl phthalate 97%
Dipentyl phthalate
Formula:
C₁₈H₂₆O₄ MW: 306.41 g/mol Boiling Pt: 342 °C (760 mmHg) Melting Pt: –55 °C Density: 1.023 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00041934 CAS Number: 131-18-0 EINECS: 205-017-9 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4885 to 1.4905 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...