2,7-Dibromofluorene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,7-Dibromo-9H-fluorene

35524-0250 35524-0050
TS35524-0250EA 94 USD
TS35524-0250 TS35524-0050
2,7-Dibromofluorene 99%
2,7-Dibromofluorene
Formula: C₁₃H₈Br₂
MW: 324.01 g/mol
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00019048
CAS Number: 16433-88-8

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 164 to 167 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR