2-(4-Methoxyphenyl)ethanol 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methoxyphenethyl alcohol

35405-0100 35405-0500
TS35405-0100EA 80 USD
TS35405-0100 TS35405-0500
2-(4-Methoxyphenyl)ethanol 96%
2-(4-Methoxyphenyl)ethanol
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Melting Pt: 28…30 °C
Flash Pt: >112 °C
MDL Number: MFCD00002900
CAS Number: 702-23-8
EINECS: 211-866-6

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5360 to 1.5380 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR