4-Ethyl-2-methoxyphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

35196-0050 35196-0250
TS35196-0050EA 38 USD
TS35196-0050 TS35196-0250
4-Ethyl-2-methoxyphenol 98%
4-Ethyl-2-methoxyphenol
Formula: C₉H₁₂O₂
Boiling Pt: 234…236 °C
Melting Pt: 15 °C
Flash Pt: 107 °C
MDL Number: MFCD00038714
CAS Number: 2785-89-9
EINECS: 220-500-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5300 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR