4-Hydroxy-3,5-dimethylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

35176-0050 35176-0250 35176-0010
TS35176-0050EA 113 USD
TS35176-0050 TS35176-0250 TS35176-0010
4-Hydroxy-3,5-dimethylbenzoic acid 98%
4-Hydroxy-3,5-dimethylbenzoic acid
Formula: C₉H₁₀O₃
Melting Pt: 221…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00016536
CAS Number: 4919-37-3

Order Now

Specification Test Results

Appearance Off-white to light beige to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 221 to 226 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR