5,5'-Methylenedisalicylic acid 95%

Supplier: Thermo Scientific Chemicals

35173-0050 35173-0250
TS35173-0050EA 56 USD
TS35173-0050 TS35173-0250
5,5'-Methylenedisalicylic acid 95%
5,5'-Methylenedisalicylic acid
Formula: C₁₅H₁₂O₆
Melting Pt: 235…242 °C
Storage Temperature: Ambient
CAS Number: 122-25-8

Order Now

Specification Test Results

Appearance Cream Powder
Infrared spectrum Conforms
Melting point 232 to 241 °C
Titration with NaOH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR