Diphenhydramine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Benzhydryloxy)-N,N-dimethylethanamine hydrochloride, Diphenhydramine HCl, 2-(Benzhydryloxy)-N,N-dimethylethylamine Hydrochloride, 2-Diphenylmethoxy-N,N-dimethylethylamine Hydrochloride

35085-0250 35085-1000
TS35085-0250EA 46 USD
TS35085-0250 TS35085-1000
Diphenhydramine hydrochloride 99%
Diphenhydramine hydrochloride
Formula: C₁₇H₂₁NO
MW: 291.82 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00012479
CAS Number: 147-24-0

Order Now

Specification Test Results

Appearance White Adhering crystalline powder or crystals
Infrared spectrum Conforms
Melting point 168 to 172 °C
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR