2-(3-Bromophenyl)ethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

35056-0010 35056-0050 35056-0250
TS35056-0010EA 49 USD
TS35056-0010 TS35056-0050 TS35056-0250
2-(3-Bromophenyl)ethanol 97%
2-(3-Bromophenyl)ethanol
Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 107…110 °C (1 mmHg)
Density: 1.478 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00191857
CAS Number: 28229-69-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5730 to 1.5750 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR