2-(2-Bromophenyl)ethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

35055-0010 35055-0050
TS35055-0010EA 37 USD
TS35055-0010 TS35055-0050
2-(2-Bromophenyl)ethanol 99%
2-(2-Bromophenyl)ethanol
Formula: C₈H₉BrO
Boiling Pt: 97 °C (.7 mmHg)
Flash Pt: >110 °C
MDL Number: MFCD00093566
CAS Number: 1074-16-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5755 to 1.5775 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR