4-Chlorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chlorobenzeneboronic acid, Dihydroxy(4-chlorophenyl)borane, (4-Chlorophenyl)boronic acid, (4-Chlorophenyl)metaboric acid

34441-0010 34441-0050 34441-0250
TS34441-0010EA 31 USD
TS34441-0010 TS34441-0050 TS34441-0250
4-Chlorophenylboronic acid 97%
4-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
Melting Pt: 259…262 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR