(±)-3-Methoxy-1,2-propanediol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Methoxy-1,2-propanediol, (±)-3-Methoxy-1,2-dihydroxy propane
33839-0010
33839-0050
33839-0250
TS33839-0010EA
29
USD
TS33839-0010
TS33839-0050
TS33839-0250
(±)-3-Methoxy-1,2-propanediol 98%
(±)-3-Methoxy-1,2-propanediol
Formula:
C₄H₁₀O₃ MW: 106.12 g/mol Boiling Pt: 133 °C (35 mmHg) Density: 1.12 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004719 CAS Number: 623-39-2 EINECS: 210-791-6 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4420 to 1.4450 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...