4-Fluorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Fluorophenyl)metaboric acid, 4-Fluorobenzeneboronic acid, Dihydroxy(4-fluorophenyl)borane

33410-0010 33410-0050 33410-0250
TS33410-0010EA 21 USD
TS33410-0010 TS33410-0050 TS33410-0250
4-Fluorophenylboronic acid 97%
4-Fluorophenylboronic acid
Formula: C₆H₆BFO₂
MW: 139.92 g/mol
Melting Pt: 255…259 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039136
CAS Number: 1765-93-1

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR