5-Bromo-2-hydroxybenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Bromo-2-(hydroxymethyl)phenol, (5-Bromo-2-hydroxyphenyl)methanol

33148-0050 33148-0250
TS33148-0050EA 89 USD
TS33148-0050 TS33148-0250
5-Bromo-2-hydroxybenzyl alcohol 98%
5-Bromo-2-hydroxybenzyl alcohol
Formula: C₇H₇BrO₂
MW: 203.04 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 106…111 °C
MDL Number: MFCD00004618
CAS Number: 2316-64-5
EINECS: 219-026-0
Merck Index: 12,01460

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 106 to 111 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR