1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3,6,8-Pyrenetetrasulphonic acid tetrasodium salt

32556-0010 32556-0050
TS32556-0010EA 93 USD
TS32556-0010 TS32556-0050
1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt 95%
1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt
Formula: C₁₆H₆Na₄O₁₂S₄
MW: 610.411 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263184
CAS Number: 59572-10-0

Order Now

Specification Test Results

Appearance White to yellow to green Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR