α,α,α-Trifluoro-m-toluoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride
31294-0050
31294-0250
TS31294-0050EA
30
USD
TS31294-0050
TS31294-0250
α,α,α-Trifluoro-m-toluoyl chloride 98%
α,α,α-Trifluoro-m-toluoyl chloride
Formula:
C₈H₄ClF₃O Boiling Pt: 184…186 °C (750 mmHg) Density: 1.38 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000680 CAS Number: 2251-65-2 EINECS: 218-844-5 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4760 to 1.4780 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...