α,α,α-Trifluoro-p-xylene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Trifluoromethyl)toulene, 1-Methyl-4-(trifluoromethyl)benzene, 4-Methylbenzotrifluoride

30891-0010 30891-0050
TS30891-0010EA 50 USD
TS30891-0010 TS30891-0050
α,α,α-Trifluoro-p-xylene 98%
α,α,α-Trifluoro-p-xylene
Formula: C₈H₇F₃
MW: 160.14 g/mol
Boiling Pt: 129 °C (760 mmHg)
Density: 1.144 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00075476
CAS Number: 6140-17-6
EINECS: 228-124-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR