1-Bromo-3,5-bis(trifluoromethyl)benzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Bis(trifluoromethyl)-1-bromobenzene, 1-Bromo-3,5-bis(trifluoromethyl)benzene, 5-Bromo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Bis(trifluoromethyl)-5-bromobenzene, 3,5-Bis(trifluoromethyl)bromobenzene, 5-Bromo-α,α,α,α',α',α'-Hexafluoro-m-xylene
30770-0100
30770-0500
30770-2500
TS30770-0100EA
52
USD
TS30770-0100
TS30770-0500
TS30770-2500
1-Bromo-3,5-bis(trifluoromethyl)benzene 99%
1-Bromo-3,5-bis(trifluoromethyl)benzene
Formula:
C₈H₃BrF₆ Boiling Pt: 154 °C Melting Pt: –16 °C Density: 1.71 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000381 CAS Number: 328-70-1 EINECS: 206-334-5 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4260 to 1.4280 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...