Methyl-4-(bromomethyl)benzoate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl 4-(bromomethyl)benzoate
30610-0250
30610-0050
30610-1000
TS30610-0250EA
141
USD
TS30610-0250
TS30610-0050
TS30610-1000
Methyl-4-(bromomethyl)benzoate 98%
Methyl 4-(bromomethyl)benzoate
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 130…135 °C (2 mmHg) Melting Pt: 53…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00032453 CAS Number: 2417-72-3 UN: 3261 ADR: 8 II |
Specification Test Results
Appearance | White to yellow Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 52 to 57 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...