4-Bromophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

30441-0050 30441-1000 30441-5000
TS30441-0050EA 36 USD
TS30441-0050 TS30441-1000 TS30441-5000
4-Bromophenol 97%
4-Bromophenol
Formula: C₆H₅BrO
MW: 173.01 g/mol
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002313
CAS Number: 106-41-2
EINECS: 203-394-4
Merck Index: 13,01413

Order Now

Specification Test Results

Appearance White to pink to light brown Crystalline powder or crystals or solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR