α-Bromo-2,6-difluorotoluene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
30357-0050
30357-0250
TS30357-0050EA
56
USD
TS30357-0050
TS30357-0250
α-Bromo-2,6-difluorotoluene 97%
α-Bromo-2,6-difluorotoluene
Formula:
C₇H₅BrF₂ Melting Pt: 51…55 °C Flash Pt: >109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000329 CAS Number: 85118-00-9 EINECS: 285-652-6 UN: 1759 ADR: 8,III |
Specification Test Results
Appearance | White to light beige Crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 51 to 55 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...