α,α,α-Trifluoro-p-toluoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Trifluoromethyl)benzoyl chloride, 4-(Trifluoromethyl)benzoylchloride, 4-(Trifluoromethyl)benzene-1-carbonyl chloride

30342-0050 30342-0010 30342-0250
TS30342-0050EA 64 USD
TS30342-0050 TS30342-0010 TS30342-0250
α,α,α-Trifluoro-p-toluoyl chloride 97%
α,α,α-Trifluoro-p-toluoyl chloride
Formula: C₈H₄ClF₃O
Boiling Pt: 78…79 °C (16 mmHg)
Melting Pt: –3 °C
Density: 1.4 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00000694
CAS Number: 329-15-7
EINECS: 206-342-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4755 to 1.4775 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR