4-(Trifluoromethoxy)aniline 99%

Supplier: Thermo Scientific Chemicals
Danger

29764-0025 29764-0250
TS29764-0025EA 23 USD
TS29764-0025 TS29764-0250
4-(Trifluoromethoxy)aniline 99%
4-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
Boiling Pt: 83…85 °C (15 mmHg)
Density: 1.31 g/cm³
Flash Pt: 80 °C
MDL Number: MFCD00041314
CAS Number: 461-82-5
EINECS: 207-317-5
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4640 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR