(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1R,2S)-(+)-cis-1-Amino-2-indanol, (1R,2S)-(+)-cis-1-Aminoindan-2-ol

29753-0010 29753-0100
TS29753-0010EA 48 USD
TS29753-0010 TS29753-0100
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol
Formula: C₉H₁₁NO
MW: 149.19 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00216656
CAS Number: 136030-00-7

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 117 to 121 °C
Specific optical rotation +41.5° to +47.5° (20 °C, 589 nm) (c=1, CH3OH)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR