Butyl benzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzoic acid butyl ester

29640-2500 29640-0010 29640-0050
TS29640-2500EA 41 USD
TS29640-2500 TS29640-0010 TS29640-0050
Butyl benzoate 98+%
Butyl benzoate
Formula: C₁₁H₁₄O₂
Boiling Pt: 249…251 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009439
CAS Number: 136-60-7
EINECS: 205-252-7
Merck Index: 13,01551

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4990 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR