Fullerene C60 99.9%

Supplier: Thermo Scientific Chemicals
Warning

29501-0010 29501-2500
TS29501-0010EA 232 USD
TS29501-0010 TS29501-2500
Fullerene C60 99.9%
Fullerene C60
Formula: C₆₀
Storage Temperature: Ambient
MDL Number: MFCD00151408
CAS Number: 99685-96-8
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark brown to black Powder
Infrared spectrum Conforms
HPLC ≥99.85 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR