2,6-Di-tert-butyl-4-methoxyphenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Di-tert-butyl-4-hydroxyanisole

29416-0250 29416-1000
TS29416-0250EA 57 USD
TS29416-0250 TS29416-1000
2,6-Di-tert-butyl-4-methoxyphenol 97%
2,6-Di-tert-butyl-4-methoxyphenol
Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
Boiling Pt: 135…140 °C (10 mmHg)
Melting Pt: 103…106 °C
MDL Number: MFCD00008824
CAS Number: 489-01-0
EINECS: 207-693-0

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 102 to 106 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR