3,5-Difluorobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

29347-0050 29347-0010
TS29347-0050EA 98 USD
TS29347-0050 TS29347-0010
3,5-Difluorobenzaldehyde 98+%
3,5-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 61…63 °C (43 mmHg)
Density: 1.296 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00010329
CAS Number: 32085-88-4
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear light green Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4950 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR