4-Chloro-2-fluorobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

29062-0050 29062-0250
TS29062-0050EA 181 USD
TS29062-0050 TS29062-0250
4-Chloro-2-fluorobenzaldehyde 99%
4-Chloro-2-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00143282
CAS Number: 61072-56-8

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder or crystals
Infrared spectrum Conforms
Melting point 57 to 61 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR